Läs mer
Naftopidil, TRC
Produktkod. 30466436
Change view
Kvantitet:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466436 500 mg
30471528 50 mg
30480871 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30466436 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN213501500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57149-07-2
Molekylformel C24 H28 N2 O3
Molekylvikt (g/mol) 392.49
IUPAC-namn 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol
LEDER COc1ccccc1N2CCN(CC(O)COc3cccc4ccccc34)CC2
Specifikationer
Kemiskt namn eller material Naftopidil
Kvantitet 500 mg
InChI formel InChI=1S/C24H28N2O3/c1-28-24-11-5-4-10-22(24)26-15-13-25(14-16-26)17-20(27)18-29-23-12-6-8-19-7-2-3-9-21(19)23/h2-12,20,27H,13-18H2,1H3
Formel vikt 392.21