Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.
9147.50 SEK - 26980.97 SEK
Specifications
Certifications/Compliance | Comply with FDA Reg. 177.1520 and USP Class VI |
---|---|
Type | Closed-dome Tank |
Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10050461
|
Thermo Scientific™
2650-0020 |
75 L |
9147.50 SEK
Each |
In Stock
Log in to see stock. |
|||||
10338990
|
Thermo Scientific™
2650-0030 |
115 L |
12934.30 SEK
Each |
In Stock
Log in to see stock. |
|||||
10469970
|
Thermo Scientific™
2650-0055 |
210 L |
15916.41 SEK
Each |
In Stock
Log in to see stock. |
|||||
10595971
|
Thermo Scientific™
2650-0100 |
378.5 L |
26980.97 SEK
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:- Tanks may be fitted with spigots
- Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
Warranty: 90 days
Specifications
Comply with FDA Reg. 177.1520 and USP Class VI | |
Closed-dome Tank | |
Polypropylene |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.