Läs mer
Thermo Scientific™ Nalgene™ L900 Liquid Detergent
Beskrivning
Thoroughly clean all plastic labware, especially polycarbonate, with Thermo Scientific™ Nalgene™ L900 Liquid Detergent. This environmentally friendly formula will not cause crazing, stress cracking or clouding of plastic when used as directed.
- For hand or machine use
- Biodegradable
- Non-toxic
- Low foaming
- Specially formulated for Nalgene plastic labware
Specifikationer
Specifikationer
| Beskrivning | L900 Liquid Detergent; L-900 Liquid Detergent *DO NOT FREEZE* 4 Liter |
| Kvantitet | 4 Pk. |
| Volym (engelska) | 1.06 gal. |
| Volym (metrisk) | 4 l |