Learn More
Thermo Scientific™ Nalgene™ Silicone Sanitary Gaskets for Nalgene™ Carboys
Protect against leakage, breakage and contamination with these platinum-cured silicone gaskets, ensuring the highest degree of purity for biopharmaceutical applications.
65.97 SEK - 1740.35 SEK
Specifications
Certifications/Compliance Meets USP Class VI requirements
Description Silicone Sanitary Gaskets
No. per Case 6
No. per Pack 1
View More Specs
Products 6
Product Code Brand Description Closure Size Price Quantity & Availability  
Product Code Brand Description Closure Size Price Quantity & Availability  
11339724 View Documents Thermo Scientific™
2672-0075
Silicone Sanitary Gaskets Ferrule size 0.75 in. (19 mm) 529.53 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
11-10-2019
Add to basket
15941501 View Documents Thermo Scientific™
2672-0075
Silicone Sanitary Gaskets Ferrule size 0.75 in. (19 mm) 529.53 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
11-10-2019
Add to basket
10355701 View Documents Thermo Scientific™
2672-0150
Silicone Sanitary Gaskets Ferrule size 1.5 in. (38 mm) 65.97 SEK
Each
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-10-2019
Add to basket
11349724 View Documents Thermo Scientific™
2672-0150
Silicone Sanitary Gaskets Ferrule size 1.5 in. (38 mm) 395.82 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
11-10-2019
Add to basket
10641703 View Documents Thermo Scientific™
2672-0300
Silicone Sanitary Gaskets Ferrule size 3 in. (76 mm) 1740.35 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
11-10-2019
Add to basket
10172201 View Documents Thermo Scientific™
2672-0075
Silicone Sanitary Gaskets Ferrule size 0.75 in. (19 mm) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Autoclavable
  • Use with Nalgene™ Carboys (Cat. Nos. 2261, 2630, and 2640)
Warranty: 90 days
Specifications
Specifications
Meets USP Class VI requirements
Silicone Sanitary Gaskets
6
1
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.