missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Nalidixic Acid Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175301
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL Nalidixic Acid produces 37% inhibition in a competitive ELISA employing Nalidixic Acid polyclonal antibody.
Nalidixic Acid is a synthetic quinolone and antibacterial agent with urinary tract antiseptic activity. Nalidixic acid concentrates in the renal tubules and bladder and exerts its local antibacterial actions by interference of DNA gyrase activity, thereby inhibiting DNA synthesis during bacterial replication in a dose-dependent manner. Nalidixic acid is active against most gram-negative organisms that cause urinary tract infections. [National Center for Biotechnology Information. PubChem Compound Database; CID=4421].
Specifikationer
| Nalidixic Acid | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.1 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Unconjugated | |
| C12H12N2O3; Nalidixate; Nalidixin; Nevigramon; Unaserus | |
| Nalidixic Acid-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering