missing translation for 'onlineSavingsMsg'
Läs mer

Naloxonhydrokloriddihydrat 1,0 mg/ml i metanol (som vattenfri bas), LoGiCal

Product Code. 30458567
Change view
Click to view available options
Kvantitet:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30458567 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458567 Supplier LoGiCal Supplier No. LGCAMP0272.0001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal levererar certifierade referensmaterial som du kan lita på som prydliga, inbyggda lösningar och stabila isotopmärkta lösningar, med noggrann dokumentation och expertstöd. Från rättsmedicinska till kliniska laboratorier, välj LoGiCal för självförtroende, efterlevnad och analytisk excellens.

Naloxonhydrokloriddihydrat 1,0 mg/ml i metanol (som vattenfri bas)
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Naloxone hydrochloride dihydrate
CAS 51481-60-8
Analyt- eller komponentnamn Naloxone hydrochloride dihydrate
Frakt skick Ispåse (-20 °C)
Molekylformel C19 H21 N O4 . Cl H . 2 H2 O
Certifikat och efterlevnad ISO 17034
Synonym Naloxone Hydrochloride Dihydrate, Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propenyl)-, hydrochloride, dihydrate, (5 alpha)-
InChI formel InChI=1 S/C19H21NO4.ClH0.2H2O/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11;;;/h2-4,14,17,21,23 H,1,5-10H2;1 H;2*1H2/t14-,17+,18+,19-;;;/m1.../s1
LEDER O.O.Cl.Oc1ccc2C[C@H]3 N(CC=C)CC[C@@]45 [C@@H](Oc1c24)C(=O)CC[C@@]35 O
IUPAC-namn (4 R,4 AS,7 aR,12 bS)-4 A,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7 A,13-hexahydro-1 H-4,12-metanobenzofuro[3,2-e]isokinolin-7-one; dihydrat; hydroklorid
Lösningsmedel eller matris Metanol
Industry Type Kriminalteknik
Molekylvikt (g/mol) 399.87
Kvantitet 1 ml
Formel vikt 399,1449 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.