Läs mer
Naloxone, TRC
Produktkod. 30010685
Change view
Kvantitet:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010685 100 mg
30007159 1 mg
30004154 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN284995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 465-65-6
Molekylformel C19 H21 N O4
Molekylvikt (g/mol) 327.37
Synonym Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propen-1-yl)-, (5α)-, Morphinan-6-one, 17-allyl-4,5α-epoxy-3,14-dihydroxy- (8CI), Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propenyl)-, (5α)- (9CI), Normorphinone, N-allyl-7,8-dihydro-14-hydroxy- (7CI), (-)-Naloxone, 12-Allyl-7,7a,8,9-tetrahydro-3,7a-dihydroxy-4aH-8,9c-iminoethanophenanthro[4,5-bcd]furan-5(6H)-one, NSC 70413, Naloxone, l-Naloxone
IUPAC-namn (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one
LEDER Oc1ccc2C[C@H]3N(CC=C)CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O
Specifikationer
Kemiskt namn eller material Naloxone
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C19H21NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,14,17,21,23H,1,5-10H2/t14-,17+,18+,19-/m1/s1
Formel vikt 327.1471
Procent renhet >95%