missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Ndelta-trityl-L-glutamine, 98%

Brand:  Alfa Aesar™ H63549.14

Additional Details : CAS Number : 132388-60-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15438366

  • 4998.50 SEK / 25g


Estimated Shipment
23-10-2020
Add to basket

Specifications

Chemical Identifiers

132388-60-4
522.601
MYOAIKMOWHPBQS-NDEPHWFRSA-N
10962399
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)NC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)O
C32H30N2O5
MFCD00144845
z-gln trt-oh, cbz-gln trt-oh, z-l-gln trt-oh, n-cbz-n'-trityl-l-glutamine, s-2-benzyloxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-benzyloxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem15365, 2s-2-benzyloxycarbonylamino-5-oxo-5-tritylamino pentanoic acid
(2S)-5-oxo-2-(phenylmethoxycarbonylamino)-5-(tritylamino)pentanoic acid

Specifications

Nalpha-Benzyloxycarbonyl-Ndelta-trityl-L-glutamine
C32H30N2O5
z-gln trt-oh, cbz-gln trt-oh, z-l-gln trt-oh, n-cbz-n'-trityl-l-glutamine, s-2-benzyloxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-benzyloxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem15365, 2s-2-benzyloxycarbonylamino-5-oxo-5-tritylamino pentanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)NC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)O
522.601
522.59
98%
132388-60-4
MFCD00144845
MYOAIKMOWHPBQS-NDEPHWFRSA-N
(2S)-5-oxo-2-(phenylmethoxycarbonylamino)-5-(tritylamino)pentanoic acid
10962399
Solid
25g
Videos
SDS Documents