Learn More
Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine, 98%, Thermo Scientific™
Catalog Number p-7054142
Click to view available options
Quantity:
1 g
5 g
76477-42-4
C19H28N2O6
380.44
MFCD00038262
BDHUTRNYBGWPBL-GGYSOQFKNA-N
boc-d-lys z-oh, n-boc-n'-cbz-d-lysine, boc-l-lys z-oh, boc-n-epsilon-z-d-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-d-lysine, 2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, ambotzbaa1358, n-alpha-boc-n-delta-z-d-lysine, boc-d-lys z, boc-d-lysine cbz
7269335
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
76477-42-4
C19H28N2O6
380.44
MFCD00038262
BDHUTRNYBGWPBL-GGYSOQFKNA-N
boc-d-lys z-oh, n-boc-n'-cbz-d-lysine, boc-l-lys z-oh, boc-n-epsilon-z-d-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-d-lysine, 2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, ambotzbaa1358, n-alpha-boc-n-delta-z-d-lysine, boc-d-lys z, boc-d-lysine cbz
7269335
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Formula Weight 380.44
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine