missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine, 95%, Thermo Scientific™

Catalog Number p-7054742
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
16965-06-3
C13H21F3N2O5
342.32
MFCD00037104
DEIYNDIFGSDDCY-SVGMAFHSNA-N
boc-lys tfa-oh, s-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, boc-l-lys tfa-oh, 2s-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, ambotzbaa5790, n-a-boc-n-epsilon-trifluoroacetyl-l-lysine, n-a-boc-n-e-trifluoroacetyl-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, n∼2∼-tert-butoxycarbonyl-n∼6∼-trifluoroacetyl-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-trifluoroacetamido hexanoic acid
7009540
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
This item is not returnable. View return policy
16965-06-3
C13H21F3N2O5
342.32
MFCD00037104
DEIYNDIFGSDDCY-SVGMAFHSNA-N
boc-lys tfa-oh, s-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, boc-l-lys tfa-oh, 2s-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, ambotzbaa5790, n-a-boc-n-epsilon-trifluoroacetyl-l-lysine, n-a-boc-n-e-trifluoroacetyl-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, n∼2∼-tert-butoxycarbonyl-n∼6∼-trifluoroacetyl-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-trifluoroacetamido hexanoic acid
7009540
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N alpha-Boc-N epsilon-trifluoroacetyl-L-lysine, is used as a fine chemical intermediate.

Solubility
Soluble in DMSO.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Solubility Information Soluble in DMSO.
Formula Weight 342.31
Percent Purity 95%
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'