Learn More
Alfa Aesar™ Nalpha-Boc-Nomega-(p-toluenesulfonyl)-L-arginine, 95%
Brand:  Alfa Aesar™ H62953.14
Additional Details : CAS Number : 13836-37-8 Weight : 0.02500kg
Quantity 25g
 View more versions of this product
Product Code. 15459398
  • 2159.69 SEK / 25g
Estimated Shipment date
from Supplier 02-02-2018
Add to basket
Specifications
Chemical Identifiers
Nalpha-Boc-Nomega-(p-toluenesulfonyl)-L-arginine
428.5
2683538
13836-37-8
MFCD00065557
Boc-Arg(Tos)-OH; Nα-Boc-Nω-tosyl-L-arginine
Specifications
Nalpha-Boc-Nomega-(p-toluenesulfonyl)-L-arginine
Solid
428.5
2683538
≈90°C
13836-37-8
95%
MFCD00065557
Boc-Arg(Tos)-OH; Nα-Boc-Nω-tosyl-L-arginine
25g