missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 95%

Brand:  Alfa Aesar™ H63907.14

Additional Details : CAS Number : 98930-01-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15448576

  • 1590.85 SEK / 25g


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

98930-01-9
608.71
LKGHIEITYHYVED-SANMLTNESA-N
3010428
C31H36N4O7S
MFCD00065616
Fmoc-Arg(Mtr)-OH

Specifications

Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine
95%
MFCD00065616
LKGHIEITYHYVED-SANMLTNESA-N
(2S)-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
3010428
Powder
25g
98930-01-9
C31H36N4O7S
Fmoc-Arg(Mtr)-OH
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N)C)C)OC
608.71
608.71
White
Moisture Sensitive
Videos
SDS