Läs mer
NAlpha,NEpsilon-Bis-boc-L-lysine tert-Butyl Ester, TRC
Produktkod. 30344553
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30344553 1 g
1 options
Produktkod. 30344553 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6570101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Na, Ne-Bis-boc-L-lysine tert-Butyl Ester
CAS 97347-28-9
Molekylformel C20 H38 N2 O6
Kvantitet 1 g
Synonym Na,Ne-Bis-Boc-L-Lysine Tert-Butyl Ester
InChI formel InChI=1S/C20H38N2O6/c1-18(2,3)26-15(23)14(22-17(25)28-20(7,8)9)12-10-11-13-21-16(24)27-19(4,5)6/h14H,10-13H2,1-9H3,(H,21,24)(H,22,25)/t14-/m0/s1
LEDER CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)OC(C)(C)C
IUPAC-namn tert-butyl (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
Molekylvikt (g/mol) 402.53
Formel vikt 402.273