Läs mer
Nalpha-Z-L-Lysine Benzyl Ester Benzenesulfonate Salt, TRC
Produktkod. 30476978
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30476978 5 g
1 options
Produktkod. 30476978 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4885155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nalpha-Z-L-Lysine Benzyl Ester Benzenesulfonate Salt
CAS 68973-36-4
Molekylformel C21H26N2O4.C6H6O3S
Kvantitet 5 g
Synonym N2-[(Phenylmethoxy)carbonyl]-L-lysine Phenylmethyl Ester Benzenesulfonate, N2-Carboxy-L-lysine Dibenzyl Ester Benzenesulfonate
InChI formel InChI=1S/C21H26N2O4.C6H6O3S/c22-14-8-7-13-19(20(24)26-15-17-9-3-1-4-10-17)23-21(25)27-16-18-11-5-2-6-12-18;7-10(8,9)6-4-2-1-3-5-6/h1-6,9-12,19H,7-8,13-16,22H2,(H,23,25);1-5H,(H,7,8,9)/t19-;/m0./s1
LEDER O=C(OCC1=CC=CC=C1)[C@@H](NC(OCC2=CC=CC=C2)=O)CCCCN.OS(C3=CC=CC=C3)(=O)=O
IUPAC-namn benzyl ((benzyloxy)carbonyl)-L-lysinate benzenesulfonate
Molekylvikt (g/mol) 528.62
Formel vikt 528.19