Läs mer
Nandrolone, 1mg/ml in Methanol, Lipomed
Produktkod. 30459198
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30459198 1 ml
1 options
Produktkod. 30459198 Leverantör Lipomed Leverantörsnummer LPMTES8891LM
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Nandrolone, 1mg/ml in Methanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nandrolone
CAS 434-22-0
Analyt- eller komponentnamn Nandrolone
Frakt skick Ice Pk. (-20°C)
Molekylformel C18 H26 O2
Koncentration 1 mg/mL
Synonym 17 beta-Hydroxyestr-4-en-3-one, Nandrolone, Nandrolone Decanoate Imp. D (EP), 19-Nortestosterone, Nandrolone Decanoate Imp. D (EP), Estr-4-en-3-one, 17-hydroxy-, (17 beta)- (9 CI), d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER C[C@]12 CC[C@H]3 [C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1 CC[C@@H]2 O
IUPAC-namn (8 R,9 S,10 R,13 S,14 S,17 S)-17-hydroxy-13-methyl-2,6,7,8,9,10,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-one
Lösningsmedel eller matris Methanol
Industry Type Pharmaceutical
Molekylvikt (g/mol) 274.4
Kvantitet 1 ml
Formel vikt 274.1933 g/mol