Learn More
Macherey-Nagel™ Nanocolor™ Nitrate 8 Tube Test
Guarantees maximum safety for the user because measurement is performed directly in the test tube. Macherey Nagel™ Nanocolor™ Nitrate 8 Tube Tests automatically perform the measurement after the cuvette is inserted into the cuvette slot.
Brand: Macherey-Nagel™ 985065
Additional Details : Weight : 0.41400kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Photometric determination
- Reaction basis based on international approved standard methods like DIN-, EN-, ISO-, EPA- and APHA
- Tests contain accurate predosed reagents that are directly ready to use
- Every test box is equipped with a color-coded label and can be identified clearly
Specifications
ISO 7890-1 | |
1 Pack | |
0.30 to 8.00mg/L NO3-N, 1.3 to 35.0mg/L NO3- | |
Nitrate 8 Tube Test |
Safety and Handling
- NANOCOLOR Nitrate 8
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : o-phosphoric acid, sulfuric acid, 2-propanol