Läs mer
Naphthalene-2-sulfonohydrazide, TRC
Produktkod. 30408878
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408878 100 mg
30428651 500 mg
30417694 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30408878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN756615100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 10151-46-9
Molekylformel C10 H10 N2 O2 S
Molekylvikt (g/mol) 222.264
Synonym (Naphthalen-2-yl)sulfonyl Hydrazine, (beta-Naphthylsulfonyl)hydrazine, 2-Naphthalenesulfonyl Hydrazide, 2-Naphthylsulfonyl Hydrazide2-Naphthylsulfonyl Hydrazine
IUPAC-namn naphthalene-2-sulfonohydrazide
LEDER NNS(=O)(=O)c1ccc2ccccc2c1
Specifikationer
Kemiskt namn eller material Naphthalene-2-sulfonohydrazide
Kvantitet 100 mg
InChI formel InChI=1S/C10H10N2O2S/c11-12-15(13,14)10-6-5-8-3-1-2-4-9(8)7-10/h1-7,12H,11H2
Formel vikt 222.046