Läs mer
Naphthol AS-MX Phosphate Disodium Salt, TRC
Produktkod. 30470912
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470912 250 mg
30466182 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30470912 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN367978250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96189-12-7
Molekylformel C19H16NNa2O5P
Molekylvikt (g/mol) 415.29
Synonym 3-Hydroxy-2-naphtho-2',4'-xylidide Phosphate Disodium Salt, N-(2,4-Dimethylphenyl)-3-(phosphonooxy)-2-naphthalenecarboxamide Disodium Salt
IUPAC-namn disodium;[3-[(2,4-dimethylphenyl)carbamoyl]naphthalen-2-yl] phosphate
LEDER [Na+].[Na+].Cc1ccc(NC(=O)c2cc3ccccc3cc2OP(=O)([O-])[O-])c(C)c1
Specifikationer
Kemiskt namn eller material Naphthol AS-MX Phosphate Disodium Salt
Kvantitet 250 mg
InChI formel InChI=1S/C19H18NO5P.2Na/c1-12-7-8-17(13(2)9-12)20-19(21)16-10-14-5-3-4-6-15(14)11-18(16)25-26(22,23)24;;/h3-11H,1-2H3,(H,20,21)(H2,22,23,24);;/q;2*+1/p-2
Formel vikt 415.0561