Läs mer
Naproxen Isopropyl Ester, Mikromol™
Produktkod. 30052653
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052653 100 mg
30051534 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052653 Leverantör Mikromol Leverantörsnummer MM0152.20
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Naproxen Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Naproxen Isopropyl Ester
CAS 156967-24-7
Analyt- eller komponentnamn Naproxen Isopropyl Ester
Frakt skick Room Temperature
Molekylformel C17 H20 O3
Certifikat och efterlevnad ISO 17025
Synonym Naproxen Isopropyl Ester, (alphaS)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid 1-methylethyl ester, 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, 1-methylethyl ester, (S)-, (+)-Naproxen 2-propyl ester, Naproxen isopropyl ester
InChI formel InChI=1S/C17H20O3/c1-11(2)20-17(18)12(3)13-5-6-15-10-16(19-4)8-7-14(15)9-13/h5-12H,1-4H3/t12-/m0/s1
LEDER COc1ccc2cc(ccc2c1)[C@H](C)C(=O)OC(C)C
IUPAC-namn propan-2-yl (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
Molekylvikt (g/mol) 272.34
Kvantitet 100 mg
Formel vikt 272.1412 g/mol