Learn More
Naratriptan 2-Carboxylic Acid, TRC
Product Code. 30480251
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Product Code. Quantity Unit Size
30480251 0.5 mg 0.5mg
1 options
Product Code. 30480251 Supplier Toronto Research Chemicals Supplier No. TRCN3787450.5MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Naratriptan 2-Carboxylic Acid
CAS 874583-40-1
Molecular Formula C18H25N3O4S
Quantity 0.5 mg
Synonym 3-(1-Methyl-4-piperidyl)-5-[2-(methylsulfamoyl)ethyl]-1H-indole-2-carboxylic Acid
InChI Formula InChI=1S/C18H25N3O4S/c1-19-26(24,25)10-7-12-3-4-15-14(11-12)16(17(20-15)18(22)23)13-5-8-21(2)9-6-13/h3-4,11,13,19-20H,5-10H2,1-2H3,(H,22,23)
SMILES CNS(=O)(=O)CCc1ccc2[nH]c(C(=O)O)c(C3CCN(C)CC3)c2c1
IUPAC Name 3-(1-methylpiperidin-4-yl)-5-[2-(methylsulfamoyl)ethyl]-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 379.47
Formula Weight 379.1566