Läs mer
Naratriptan Hydrochloride, TRC
Produktkod. 30481615
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481615 10 mg
30479122 50 mg
30464424 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN37873010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 143388-64-1
Molekylformel C17 H25 N3 O2 S . Cl H
Molekylvikt (g/mol) 371.93
Synonym Naratriptan Hydrochloride, 1H-Indole-5-ethanesulfonamide, N-methyl-3-(1-methyl-4-piperidinyl)-, hydrochloride (1:1), 1H-Indole-5-ethanesulfonamide, N-methyl-3-(1-methyl-4-piperidinyl)-, monohydrochloride (9CI), Amerge, GR 85548A, N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulfonamide hydrochloride, Naramig, Naratrex 1
IUPAC-namn N-methyl-2-[3-(1-methylpiperidin-4-yl)-1H-indol-5-yl]ethanesulfonamide;hydrochloride
LEDER Cl.CNS(=O)(=O)CCc1ccc2[nH]cc(C3CCN(C)CC3)c2c1
Specifikationer
Kemiskt namn eller material Naratriptan Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C17H25N3O2S.ClH/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;/h3-4,11-12,14,18-19H,5-10H2,1-2H3;1H
Formel vikt 371.1434
Procent renhet >95%