Läs mer
Narciclasine, TRC
Produktkod. 17909739
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17909739 5 mg
18041430 1 mg
18033530 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17909739 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3787935MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29477-83-6
Molekylformel C14 H13 N O7
Molekylvikt (g/mol) 307.2555
Synonym [1,3]Dioxolo[4,5-j]phenanthridin-6(2H)-one, 3,4,4a,5-tetrahydro-2,3,4,7-tetrahydroxy-, (2S,3R,4S,4aR)-, [1,3]Dioxolo[4,5-j]phenanthridin-6(2H)-one, 3,4,4a,5-tetrahydro-2,3,4,7-tetrahydroxy-, [2S-(2α,3β,4β,4aβ)]-, (+)-Lycoricidinol, (+)-Narciclasine, Lycoricidinol
IUPAC-namn (2S,3R,4S,4aR)-2,3,4,7-tetrahydroxy-3,4,4a,5-tetrahydro-2H-[1,3]dioxolo[4,5-j]phenanthridin-6-one
LEDER O[C@H]1C=C2[C@@H](NC(=O)c3c(O)c4OCOc4cc23)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Narciclasine
Kvantitet 5 mg
InChI formel InChI=1S/C14H13NO7/c16-6-1-5-4-2-7-13(22-3-21-7)11(18)8(4)14(20)15-9(5)12(19)10(6)17/h1-2,6,9-10,12,16-19H,3H2,(H,15,20)/t6-,9+,10+,12-/m0/s1
Formel vikt 307.0692