Läs mer
Narcotolinogendiol, TRC
Produktkod. 30136748
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136748 100 mg
30010568 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136748 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN378850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5876-41-5
Molekylformel C21H25NO7
Molekylvikt (g/mol) 403.43
Synonym 1,3-Dioxolo[4,5-g]isoquinoline, 1,2-benzenedimethanol deriv., Narcotolinediol, [S-(R*,S*)]-3,4-Dimethoxy-alpha1-(5,6,7,8-tetrahydro-4-hydroxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl)-1,2-benzenedimethanol
IUPAC-namn (5R)-5-[(R)-hydroxy-[2-(hydroxymethyl)-3,4-dimethoxyphenyl]methyl]-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-4-ol
LEDER COc1ccc([C@@H](O)[C@@H]2N(C)CCc3cc4OCOc4c(O)c23)c(CO)c1OC
Specifikationer
Kemiskt namn eller material Narcotolinogendiol
Kvantitet 100 mg
InChI formel InChI=1S/C21H25NO7/c1-22-7-6-11-8-15-21(29-10-28-15)19(25)16(11)17(22)18(24)12-4-5-14(26-2)20(27-3)13(12)9-23/h4-5,8,17-18,23-25H,6-7,9-10H2,1-3H3/t17-,18-/m1/s1
Formel vikt 403.1631