missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12173-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Several proteins have been found to be prenylated and methylated at their carboxyl-terminal ends. Prenylation was initially believed to be important only for membrane attachment. However, another role for prenylation appears to be its importance in protein-protein interactions. The only nuclear proteins known to be prenylated in mammalian cells are prelamin A- and B-type lamins. Prelamin A is farnesylated and carboxymethylated on the cysteine residue of a carboxyl-terminal CaaX motif. This post-translationally modified cysteine residue is removed from prelamin A when it is endoproteolytically processed into mature lamin A. The protein encoded by this gene binds to the prenylated prelamin A carboxyl-terminal tail domain. It may be a component of a prelamin A endoprotease complex. The encoded protein is located in the nucleus, where it partially colocalizes with the nuclear lamina. It shares limited sequence similarity with iron-only bacterial hydrogenases. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene, including one with a novel exon that is generated by RNA editing.Specifications
NARF | |
Polyclonal | |
Unconjugated | |
NARF | |
IOP2, NARF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26502, 360681, 67608 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2YDU6, Q9CYQ7, Q9UHQ1 | |
Narf | |
NARF Fusion Protein Ag2816 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |