Learn More
Buerkle™ Narrow Neck Reagent Bottles
Product Code. 31188970 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
2.5 L
Product Code. Capacity (Metric)
31188970 2.5 L
1 options
Product Code. 31188970 Supplier Buerkle™ Supplier No. 03402500
  • ONLINE EXCLUSIVE PRICE
    124.00 SEK
    115.17 SEK / Paket med 1
    Save 8.83 SEK
    7% Av
I lager
Universal bottles for storing, packing, taking samples, sampling and transport of liquids.
  • HDPE, transparent
  • PP tamper-evident screw cap, blue
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2.5 L
Color Translucent White
Material HDPE, Polypropylene
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Autoclavable Not Autoclavable
Capacity (English) 84.54 fl oz.
Product Type Reagent Bottle