missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NAT5 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895154
|
Proteintech
15807-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16885154
|
Proteintech
15807-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NAT5 is a component of N-acetyltransferase complex B (NatB). Human NatB performs cotranslational N(alpha)-terminal acetylation of methionine residues when they are followed by asparagine (Starheim et al., 2008Specifications
NAT5 | |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P61599, P61600 | |
362228, 51126, 67877 | |
NAT5 Fusion Protein Ag8541 | |
Primary | |
-20°C | |
NAA20 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
N acetyltransferase 5, NAA20, NAT3, NAT5, NatB complex subunit NAT5 | |
NAA20 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |