Läs mer
Thermo Scientific™ National Inserts for Standard-Opening Vials
Beskrivning
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
- Pulled point inserts are an economical choice for non-critical applications
Specifikationer
Specifikationer
| Beskrivning | C4012-531L |
| Material | Glass |
| Volym (metrisk) | 225 μl |
| Stil | Precision Point |
| Basstil | Conical |
| Produktlinje | National |