Learn More
National Scientific™ MicroTube™ Rack System
POA
Specifications
Description MicroTube Dimensions: 9mm dia., 46mm height
Material Polypropylene
View More Specs
Description
Description
Featuring a rack with an 8 x 12 hole-pattern on traditional 9mm centers, the entire System conforms to the SBS footprint/configuration compatible with robotic workstations, multichannel pipettors and auto-sampling instruments. An alpha-numeric, non-reversing rack and lid ensure positive sample identification. The lid, with individual condensation rings above each tube, prevents cross-contamination. The inert polypropylene tubes, rack and lid can be autoclaved, centrifuged at moderate speeds and stacked for long-term, low temperature storage of biological samples. The System is ideal for:
  • Bacterial and hybridoma cell uptake
  • Cell harvesting
  • CMV, HSV, HTLV-lll testing
  • Receptor binding assays
  • RIA/EIA serial dilution procedures
  • Recombinant DNA work
  • Pharmaceutical and human genome studies
  • Sample storage librariesRacks, with SBS footprint, feature 8 x 12 hole pattern on 9mm center. making compatible with robotic workstations & multichannel pipettors.
Specifications
Specifications
MicroTube Dimensions: 9mm dia., 46mm height