Läs mer
Thermo Scientific™ National Scientific septa for 4mL Screw-Thread Vials
Beskrivning
Thermo Scientific™ National Vial Septa are designed for use with 4mL screw-thread vials.
- PTFE disk septa are chemically inert
- PFTE/red rubber septa are economical for many GC applications
- PTFE/silicone septa are most popular for HPLC and IC applications
- Pre-slit septa are easier to pierce with thin gauge needles
Specifikationer
Specifikationer
| Beskrivning | Blue PTFE/White Silicone; Preslit |
| Material | PTFE, Silicone |
| Produktlinje | National |
| Stängningsstorlek | 13-425 |
| Produkttyp | Septa, Single Slit |