Learn More
Ark Plas Products™ Natural Polypropylene Commercial Grade Barbed Tee Connectors
Use these fittings for a wide range of applications. Ark Plas Products™ Natural Polypropylene Commercial Grade Barbed Tee Connectors are designed as a low cost alternative with high end performance.
950.00 SEK - 1340.00 SEK
Specifications
Product Type | Barbed Tee Connector |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Packaging Quantity | 25 Pack |
Shape | Tee |
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
11741956
|
Ark Plas Products™
CEA34-PR0 |
3/8 x 3/8 x 3/8 in. Barb |
1340.00 SEK
Pack of 25 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
11711956
|
Ark Plas Products™
CEA37-PR0 |
5/32 x 5/32 x 5/32 in. Barb |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Economic and reliable
- Up to 125psi without clamp
- Contain recycled runner material
- Available in Natural Nylon or Natural Polypropylene
Specifications
Barbed Tee Connector | |
Polypropylene | |
Tee |
Autoclavable | |
25 Pack | |
Natural |