Läs mer
Navtemadlin, MedChemExpress
Produktkod. 16485806
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16485806 100 mg -
16435078 10 mg 10mg
16401317 50 mg -
16437797 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16485806 Leverantör Medchem Express Leverantörsnummer HY12296
MedChemExpress Navtemadlin (AMG 232) is a potent, selective and orally available inhibitor of p53-MDM2 interaction, with an IC50 of 0.6 nM. Navtemadlin binds to MDM2 with a Kd of 0.045 nM.
Navtemadlin (AMG 232) is a potent, selective and orally available inhibitor of p53-MDM2 interaction, with an IC50 of 0.6 nM. Navtemadlin binds to MDM2 with a Kd of 0.045 nM.
Specifikationer
Kemiskt namn eller material Navtemadlin
CAS 1352066-68-2
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C28H35Cl2NO5S
Kvantitet 100 mg
Synonym AMG 232 KRT-232
Löslighetsinformation DMSO : ≥ 50 mg/mL (87.94 mM) ∣H2O : ≥ 0.1 mg/mL (0.18 mM)
LEDER O=C(O)C[C@]1(C)C(N([C@H](CS(=O)(C(C)C)=O)C(C)C)[C@H](C2=CC=C(Cl)C=C2)[C@@H](C3=CC=CC(Cl)=C3)C1)=O
Molekylvikt (g/mol) 568.55
Formel vikt 568.55
Procent renhet 96.04%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death