Läs mer
Naxagolide Hydrochloride, TRC
Produktkod. 30480114
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30480114 1 mg
1 options
Produktkod. 30480114 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3798001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Naxagolide Hydrochloride
CAS 99705-65-4
Molekylformel C15 H21 N O2 . Cl H
Kvantitet 1 mg
Synonym 2H-Naphth[1,2-b]-1,4-oxazin-9-ol','3,4,4a,5,6,10b-hexahydro-4-propyl-','hydrochloride (1:1)','(4aR,10bR)-, 2H-Naphth[1,2-b]-1,4-oxazin-9-ol','3,4,4a,5,6,10b-hexahydro-4-propyl-','hydrochloride','(4aR,10bR)- (9CI), 2H-Naphth[1,2-b]-1,4-oxazin-9-ol','3,4,4a,5,6,10b-hexahydro-4-propyl-','hydrochloride','(4aR-trans)-, L 647339, MK 458, Naxagolide hydrochloride, (4aR,10bR)-3,4,4a,5,6,10b-Hexahydro-4-propyl-2H-naphth[1,2-b]-1,4-oxazin-9-ol hydrochloride
InChI formel InChI=1S/C15H21NO2.ClH/c1-2-7-16-8-9-18-15-13-10-12(17)5-3-11(13)4-6-14(15)16;/h3,5,10,14-15,17H,2,4,6-9H2,1H3;1H/t14-,15-;/m1./s1
LEDER Cl.CCCN1CCO[C@H]2[C@H]1CCc3ccc(O)cc23
IUPAC-namn (4aR,10bR)-4-propyl-2,3,4a,5,6,10b-hexahydrobenzo[h][1,4]benzoxazin-9-ol;hydrochloride
Molekylvikt (g/mol) 283.79
Formel vikt 283.1339