Läs mer
Tocris Bioscience™ NBD-PE
Beskrivning
NBD-PE is a fluorescent label for lysosomal lipid bodies. It consists of a single tail (16:0) phospholipid derivative with an NBD fluorophore conjugated to the head group. NBD-PE can be used to quantitatively measure phospholipidosis, which is useful to screen drugs for SARS-CoV-2 repurposing. NBD-PE can also be used to label giant unilamellar vesicles (GUV).
Specifikationer
Specifikationer
| Kemiskt namn eller material | Triethylammonium (R)-2,3-bis(palmitoyloxy)propyl(2-((7-nitrobenzo[c][1,2,5 ]oxadiazol-4-yl)amino)ethyl)phosphate |
| CAS | 178119-00-1 |
| Kvantitet | 10 mg |
| Mål | Fluorescent Lipids Probes and Cell Membrane Stains |
| Molekylformel | C43H75N4O11P.C6H15N |
| Renhet | 0.95 |