missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
NCAM-1/CD56 Antibody (SPM128), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34397APC
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Rat, Zebrafish samples. It is validated for Flow Cytometry.
Specifikationer
| NCAM-1/CD56 | |
| Monoclonal | |
| APC | |
| CD56, CD56 NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is also expressed on some mesodermally derived tumors (rhabdomyosarcoma). Anti-CD56 plays an important role in the diagnosis of nodal and nasal NK/T-cell lymphomas. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| SPM128 | |
| Flow Cytometry | |
| NCAM1 | |
| Membrane preparation of a small cell lung carcinoma | |
| 0.1 mL | |
| Astrocyte Markers, Cellular Markers, Cytokine Research, Cytoskeleton Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers | |
| 4684 | |
| Human, Rat, Zebrafish | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering