Läs mer
Tocris Bioscience™ NCT 501 Hydrochloride, Tocris Bioscience™
Potent and selective ALDH1A1 inhibitor
Brand: Tocris Bioscience™ 5934/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NCT 501 hydrochloride is a potent and selective ALDH1A1 inhibitor (IC50 = 40 nM). Exhibits >1000-fold selectivity for ALDH1A1 over ALDH3A1, ALDH1B1, ALDH2, HPGD and HSD17β4. Also selective over a panel of 168 GPCRs. Blood-brain barrier permeable.Specifikationer
| 8-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]methyl]-3,7-dihydro-1,3-dimethyl-7-(3-methylbutyl)-1H-purine-2,6-dione hydrochloride | |
| 50 mg | |
| Solid | |
| Bioactive Small Molecule |
| 1307896-46-3 | |
| Soluble to 100mM in water and to 100mM in DMSO | |
| White |