missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine dicyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H62206.06

Additional Details : CAS Number : 13665-13-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497156

  • 1772.18 SEK / 5g


Estimated Shipment date
from Supplier 25-04-2019
Add to basket

Specifications

Chemical Identifiers

13665-13-9
547.737
DZDMNVANCXRHHF-UQKRIMTDSA-N
56777030
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
C30H49N3O6
MFCD00270629
z-orn boc-oh dcha, 2s-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid; dicha, dicyclohexylamine s-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoate, n5-1,1-dimethylethoxy carbonyl-n2-phenylmethoxy carbonyl-l-ornithine dicyclohexylammonium salt
N-cyclohexylcyclohexanamine;(2S)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine dicyclohexylammonium salt
C30H49N3O6
z-orn boc-oh dcha, 2s-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid; dicha, dicyclohexylamine s-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoate, n5-1,1-dimethylethoxy carbonyl-n2-phenylmethoxy carbonyl-l-ornithine dicyclohexylammonium salt
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
547.737
547.73
95%
13665-13-9
MFCD00270629
DZDMNVANCXRHHF-UQKRIMTDSA-N
N-cyclohexylcyclohexanamine;(2S)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid
56777030
Solid
5g
Videos
SDS