missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDNL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NDNL2 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825655
|
Proteintech
27488-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815655
|
Proteintech
27488-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is part of the SMC5-6 chromatin reorganizing complex and is a member of the MAGE superfamily. This is an intronless gene. Component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintece via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs).Specifications
NDNL2 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96MG7 | |
56160 | |
NDNL2 Fusion Protein Ag26663 | |
Primary | |
-20°C | |
NSMCE3 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HCA4, MAGE G1 antigen, MAGEG1, MAGEL3, Melanoma associated antigen G1, NDNL2, necdin like 2, Necdin like protein 2, NSE3, NSMCE3 | |
NSMCE3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |