Learn More
NDRG2 Mouse anti-Human, Unconjugated, Clone: 3E3F4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67191-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839695
  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The NDRG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NDRG2. This antibody recognizes human, rat, mouse, pig antigen. The NDRG2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NDRG2
Monoclonal
1000 μg/mL
Mouse
Primary
Pig, Rat, Mouse, Human
ELISA, Immunohistochemistry, Western Blot
3E3F4
Unconjugated
150 μL
57447
IgG2b