missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12184-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
NDRG4 | |
Polyclonal | |
Unconjugated | |
NDRG4 | |
BDM1, KIAA1180, NDRG family member 4, NDRG4, Protein NDRG4, SMAP 8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234593, 65009 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BTG7, Q9ULP0 | |
NDRG4 | |
NDRG4 Fusion Protein Ag3029 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |