Läs mer
Tocris Bioscience™ NDT 9513727
GREENER_CHOICE
Produktkod. 18025782
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18025782 50 mg
18013792 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18025782 Leverantör Tocris Bioscience™ Leverantörsnummer 3333/50
Potent, selective human C5a receptor negative allosteric modulator (NAM)
Potent C5a receptor (C5aR) negative allosteric modulator (NAM) (IC50 = 11.6 nM); selective for human C5aR (IC50 values for rat and mouse C5aR are > 10 μM). Exhibits inverse agonist pharmacokinetics. Blocks degranulation response in C5a-stimulated U937 cells (IC50 = 7.1 nM). Exhibits selectivity for primate and gerbil receptor over other species. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 439571-48-9
Molekylformel C36H35N3O4
Specifikationer
Kemiskt namn eller material NDT 9513727
Kvantitet 50 mg
Formel vikt 573.68
Renhet >98%
Molekylvikt (g/mol) 573.693