missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ NDUFAF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562340
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: VNDLPRALEQ IHYILKPDGV FIGAMFGGDT LYELRCSLQL AETEREGGFS PHISPFTAVN DLGHLLGRAG FNTLTVDTDE IQVNYPGMFE LMED Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes a mitochondrial protein that is associated with the matrix face of the mitochondrial inner membrane and is required for complex I assembly. A mutation in this gene results in mitochondrial complex I deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifikationer
| NDUFAF5 | |
| Polyclonal | |
| Unconjugated | |
| NDUFAF5 | |
| 2310003L22Rik; arginine-hydroxylase NDUFAF5, mitochondrial; bA526K24.2; C13H20orf7; C20orf7; C24H20orf7; dJ842G6.1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; arginine-hydroxylase NDUFAF5, mitochondrial; NADH:ubiquinone oxidoreductase complex assembly factor 5; ndufaf5; probable methyltransferase C20orf7 homolog, mitochondrial; probable methyltransferase C20orf7, mitochondrial; putative methyltransferase C20orf7, mitochondrial-like protein; putative methyltransferase NDUFAF5; RGD1309829; zgc:162919 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79133 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TEU4 | |
| NDUFAF5 | |
| Recombinant protein corresponding to Human NDUFAF5. Recombinant protein control fragment (Product #RP-99326). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering