Learn More
NDUFB10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16854824 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16854824 20 μL 20µL
16844824 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16854824 Supplier Proteintech Supplier No. 155891AP20UL
Rabbit Polyclonal Antibody
NDUFB10 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. Complex I is composed of 45 different subunits.
Specifications
Antigen NDUFB10
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFB10
Gene Accession No. O96000, Q9DCS9
Gene Alias CI PDSW, Complex I PDSW, NDUFB10, PDSW
Gene Symbols NDUFB10
Host Species Rabbit
Immunogen NDUFB10 Fusion Protein Ag7922
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4716, 681418, 68342
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG