missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NDUFS1 (Human) Recombinant Protein
Human NDUFS1 full-length ORF ( AAH30833, 1 a.a. - 727 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004719-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLRIPVRKALVGLSKSPKGCVRTTATAASNLIEVFVDGQSVMVEPGTTVLQACEKVGMQIPRFCYHERLSVAGNCRMCLVEIEKAPKVVAACAMPVMKGWNILTNSEKSKKAREGVMEFLLANHPLDCPICDQGGECDLQDQSMMFGNDRSRFLEGKRAVEDKNIGPLVKTIMTRCIQCTRCIRFASEIAGVDDLGTTGRGNDMQVGTYIEKMFMSELSGNIIDICPVGALTSKPYAFTAQPWETRKTESIDVMDAVGSNIVVSTRTGEVMRILPRMHEDINEEWISDKTRFAYDGLKRQRLTEPMVRNEKGLLTYTSWEDALSRVAGMLQSFQGKDVAAIAGGLVDAEALVALKDLLNRVDSDTLCTEEVFPTAGAGTDLRSNYLLNTTIAGVEEADVVLLVGTNPRFEAPLFNARIRKSWLHNDLKVALIGSPVDLTYTYDHLGDSPKILQDIASGSHPFSQVLKEAKKPMVVLGSSALQRNDGAAILAAVSSIAQKIRMTSGVTGDWKVMNILHRIASQVAALDLGYKPGVEAIRKNPPKVLFLLGADGGCITRQDLPKDCFIIYQGHHGDVGAPIADVILPGAAYTEKSATYVNTEGRAQQTKVAVTPPGLAREDWKIIRALSEIAGMTLPYDTLDQVRNRLEEVSPNLVRYDDIEGANYFQQANELSKLVNQQLLADPLVPPQLTIKDFYMTDSISRASQTMAKCVKAVTEGAQAVEEPSIC
Specifications
AAH30833 | |
NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NDUFS1 | |
GST |
4719 | |
Wheat germ expression system | |
10 μg | |
CI-75Kd, MGC26839, PRO1304 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |