Learn More
NEDD4L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555124
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SATNSNNHLI EPQIRRPRSL SSPTVTLSAP LEGAKDSPVR RAVKDTLSNP QSPQPSPYNS PKPQHKVTQS FLPPGWEM Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
NEDD4L Polyclonal antibody specifically detects NEDD4L in Human samples. It is validated for Immunohistochemistry.Specifications
NEDD4L | |
Polyclonal | |
Unconjugated | |
NEDD4L | |
1300012C07Rik; E3 ubiquitin-protein ligase NEDD4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; hNEDD4-2; KIAA0439; NEDD4; NEDD4 like E3 ubiquitin protein ligase; NEDD4.2; Nedd4-2; nedd4b; NEDD4L; NEDL3; neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4b; neural precursor cell expressed, developmentally down-regulated gene 4-like; RSP5; ubiquitin-protein ligase Nedd4-2; ubiquitin-protein ligase Rsp5; Unknown (protein for IMAGE:7986262) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23327 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PU5 | |
NEDD4L | |
Recombinant protein corresponding to Human NEDD4L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only