missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NEDD8 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836514
|
Proteintech
16777-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826514
|
Proteintech
16777-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NEDD8 is a ubiquitin-like protein which plays an important role in cell cycle control and embryogenesis. Covalent attachment to its substrates requires prior activation by the E1 complex UBE1C-APPBP1 and linkage to the E2 enzyme UBE2M. Attachment of NEDD8 to cullins activates their associated E3 ubiquitin ligase activity, and thus promotes polyubiquitination and proteasomal degradation of cyclins and other regulatory proteins.Specifications
NEDD8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P29595, Q15843, Q71UE8 | |
18002, 25490, 4738 | |
NEDD8 Fusion Protein Ag10154 | |
Primary | |
-20°C | |
NEDD8 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
cul nedd8, Nedd 8, NEDD8, Neddylin, Ubiquitin like protein Nedd8 | |
Nedd8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |