missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16777-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
NEDD8 is a ubiquitin-like protein which plays an important role in cell cycle control and embryogenesis. Covalent attachment to its substrates requires prior activation by the E1 complex UBE1C-APPBP1 and linkage to the E2 enzyme UBE2M. Attachment of NEDD8 to cullins activates their associated E3 ubiquitin ligase activity, and thus promotes polyubiquitination and proteasomal degradation of cyclins and other regulatory proteins.Specifications
NEDD8 | |
Polyclonal | |
Unconjugated | |
NEDD8 | |
cul nedd8, Nedd 8, NEDD8, Neddylin, Ubiquitin like protein Nedd8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18002, 25490, 4738 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29595, Q15843, Q71UE8 | |
Nedd8 | |
NEDD8 Fusion Protein Ag10154 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |