Läs mer
Neem Oil, TRC
Produktkod. 30468997
Change view
Kvantitet:
1 kg
Produktkod. Kvantitet
30468997 1 kg
1 options
Produktkod. 30468997 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3889201KG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Neem Oil
CAS 8002-65-1
Kvantitet 1 kg
Synonym Azadirachta indica oil, Azadirachta indica seed oil, Margosa oil, Margosa seed oil, Melia azadirachta oil, Melia azadirachta seed oil, Neem Indica, Neem seed oil