Learn More
Nefamide, TRC
Product Code. 30481852
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481852 10 mg 10mg
30471928 100 mg -
30482350 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481852 Supplier Toronto Research Chemicals Supplier No. TRCN38911010MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24833-47-4
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.32
Synonym 2-Benzoyl-N-(2-hydroxyethyl)-N-methylbenzamide, o-Benzoyl-N-(2-hydroxyethyl)-N-methylbenzamide
IUPAC Name 2-benzoyl-N-(2-hydroxyethyl)-N-methylbenzamide
SMILES CN(CCO)C(=O)c1ccccc1C(=O)c2ccccc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H17NO3/c1-18(11-12-19)17(21)15-10-6-5-9-14(15)16(20)13-7-3-2-4-8-13/h2-10,19H,11-12H2,1H3
Formula Weight 283.1208
Chemical Name or Material Nefamide