Läs mer
Nefazodone Hydrochloride, TRC
Produktkod. 30474424
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474424 100 mg
30474948 50 mg
30477358 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474424 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN389100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82752-99-6
Molekylvikt (g/mol) 506.47
Synonym 3H-1,2,4-Triazol-3-one, 2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl-2,4-dihydro-4-(2-phenoxyethyl)-, hydrochloride (1:1)3H-1,2,4-Triazol-3-one, 2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl-2,4-dihydro-4-(2-phenoxyethyl)-, monohydrochloride (9CI), BMY 13754, BMY 13754-1, Dutonin, MJ 13754-1, Nefazodone hydrochloride, Serazone, Serzone
IUPAC-namn 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride
LEDER Cl.CCC1=NN(CCCN2CCN(CC2)c3cccc(Cl)c3)C(=O)N1CCOc4ccccc4
Specifikationer
Kemiskt namn eller material Nefazodone Hydrochloride
Molekylformel C25 H32 Cl N5 O2 . Cl H
Kvantitet 100 mg
InChI formel InChI=1S/C25H32ClN5O2.ClH/c1-2-24-27-31(25(32)30(24)18-19-33-23-10-4-3-5-11-23)13-7-12-28-14-16-29(17-15-28)22-9-6-8-21(26)20-22;/h3-6,8-11,20H,2,7,12-19H2,1H3;1H
Formel vikt 505.2011