Läs mer
(+)-Nefopam Hydrochloride, TRC
Produktkod. 30478449
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478449 50 mg
30473110 500 mg
30473243 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30478449 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN38574550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53648-31-0
Molekylformel C17H19NO . HCl
Molekylvikt (g/mol) 289.8
Synonym (S)-3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, (1S)-3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, S-Nefopam Hydrochloride
IUPAC-namn (1S)-5-methyl-1-phenyl-1,3,4,6-tetrahydro-2,5-benzoxazocine;hydrochloride
LEDER Cl.CN1CCO[C@@H](c2ccccc2)c3ccccc3C1
Specifikationer
Kemiskt namn eller material (+)-Nefopam Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C17H19NO.ClH/c1-18-11-12-19-17(14-7-3-2-4-8-14)16-10-6-5-9-15(16)13-18;/h2-10,17H,11-13H2,1H3;1H/t17-;/m0./s1
Formel vikt 289.1233